2-Amino-3-ethylquinoline hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-3-ethyl-1-azanaphthalene hydrochloride, 3-Ethylquinolin-2-amine hydrochloride

OR307520-1G OR307520-5G
APOSOR307520-1GEA 301 EUR
APOSOR307520-1G APOSOR307520-5G
2-Amino-3-ethylquinoline hydrochloride
2-Amino-3-ethylquinoline hydrochloride
Formel: C₁₁H₁₂N₂·ClH
Molecular Weight: 208,687 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10703338
CAS-Nummer: 1170053-53-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR