cis-2-Aminomethyl-cyclohexylamine dihydrochloride

Lieferant: Apollo Scientific

OR307557-1G OR307557-250MG
APOSOR307557-1GEA 1010 EUR
APOSOR307557-1G APOSOR307557-250MG
cis-2-Aminomethyl-cyclohexylamine dihydrochloride
cis-2-Aminomethyl-cyclohexylamine dihydrochloride
Formel: C₇H₁₆N₂·2ClH
Molecular Weight: 201,137 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11052861
CAS-Nummer: 1212366-42-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR