cis-6-Aminomethyl-cyclohex-3-enylamine dihydrochloride

Lieferant: Apollo Scientific

OR307558-1G OR307558-250MG
APOSOR307558-1GEA 1100 EUR
APOSOR307558-1G APOSOR307558-250MG
cis-6-Aminomethyl-cyclohex-3-enylamine dihydrochloride
cis-6-Aminomethyl-cyclohex-3-enylamine dihydrochloride
Formel: C₇H₁₄N₂·2ClH
Molecular Weight: 199,121 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11052862
CAS-Nummer: 1212102-19-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR