3-tert-Butoxycarbonylamino-hexanoic acid

Lieferant: Apollo Scientific

OR307568-1G OR307568-250MG
APOSOR307568-1GEA 648 EUR
APOSOR307568-1G APOSOR307568-250MG
3-tert-Butoxycarbonylamino-hexanoic acid
3-tert-Butoxycarbonylamino-hexanoic acid
Formel: C₁₁H₂₁NO₄
Molecular Weight: 231,289 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02258128
CAS-Nummer: 282524-95-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR