cis-2-Hydroxymethyl-1-methyl-1-cyclohexylamine hydrochloride
Lieferant: Apollo Scientific
OR307596-1G
OR307596-250MG
APOSOR307596-1GEA
739
EUR
APOSOR307596-1G
APOSOR307596-250MG
cis-2-Hydroxymethyl-1-methyl-1-cyclohexylamine hydrochloride
cis-2-Hydroxymethyl-1-methyl-1-cyclohexylamine hydrochloride
Formel:
C₈H₁₇NO·ClH Molecular Weight: 179,688 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07366579 CAS-Nummer: 1212253-95-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...