3-Amino-3-cyclohexyl-propan-1-ol
Lieferant: Apollo Scientific
OR307622-1G
OR307622-250MG
APOSOR307622-1GEA
574
EUR
APOSOR307622-1G
APOSOR307622-250MG
3-Amino-3-cyclohexyl-propan-1-ol
3-Amino-3-cyclohexyl-propan-1-ol
Formel:
C₉H₁₉NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD03412684 CAS-Nummer: 109647-95-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...