1-(1-Amino-ethyl)-naphthalen-2-ol hydrochloride

Lieferant: Apollo Scientific

OR307641-1G OR307641-250MG
APOSOR307641-1GEA 549 EUR
APOSOR307641-1G APOSOR307641-250MG
1-(1-Amino-ethyl)-naphthalen-2-ol hydrochloride
1-(1-Amino-ethyl)-naphthalen-2-ol hydrochloride
Formel: C₁₂H₁₃NO·ClH
Molecular Weight: 223.699 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08437528
CAS-Nummer: 915781-01-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR