1-(1-Amino-2-methyl-propyl)naphthalen-2-ol hydrochloride

Lieferant: Apollo Scientific

OR307643-1G OR307643-250MG
APOSOR307643-1GEA 549 EUR
APOSOR307643-1G APOSOR307643-250MG
1-(1-Amino-2-methyl-propyl)naphthalen-2-ol hydrochloride
1-(1-Amino-2-methyl-propyl)naphthalen-2-ol hydrochloride
Formel: C₁₄H₁₇NO·ClH
Molecular Weight: 251.752 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08437530
CAS-Nummer: 915781-03-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR