(1S,2R)-2-Amino-cyclohex-3-enecarboxylic acid hydrochloride
Lieferant: Apollo Scientific
OR307646-1G
OR307646-250MG
APOSOR307646-1GEA
1300
EUR
APOSOR307646-1G
APOSOR307646-250MG
(1S,2R)-2-Amino-cyclohex-3-enecarboxylic acid hydrochloride
(1S,2R)-2-Amino-cyclohex-3-enecarboxylic acid hydrochloride
Formel:
C₇H₁₁NO₂·ClH Molecular Weight: 177.629 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04112726 CAS-Nummer: 132487-40-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...