trans-tert-Butyl 3-(3-bromophenyl)-aziridine-2-carboxylate

Lieferant: Apollo Scientific

OR307705-1G OR307705-250MG
APOSOR307705-1GEA 509 EUR
APOSOR307705-1G APOSOR307705-250MG
trans-tert-Butyl 3-(3-bromophenyl)-aziridine-2-carboxylate
trans-tert-Butyl 3-(3-bromophenyl)-aziridine-2-carboxylate
Formel: C₁₃H₁₆BrNO₂
Molecular Weight: 298,176 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26936316
CAS-Nummer: 1980008-30-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR