cis-tert-Butyl 3-(4-methoxy-3-nitrophenyl)-aziridine-2-carboxylate

Lieferant: Apollo Scientific

OR307707-1G OR307707-250MG
APOSOR307707-1GEA 509 EUR
APOSOR307707-1G APOSOR307707-250MG
cis-tert-Butyl 3-(4-methoxy-3-nitrophenyl)-aziridine-2-carboxylate
cis-tert-Butyl 3-(4-methoxy-3-nitrophenyl)-aziridine-2-carboxylate
Formel: C₁₄H₁₈N₂O₅
Molecular Weight: 294,303 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26936318
CAS-Nummer: 1980007-31-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR