1-(3,4-Dimethoxyphenyl)cyclopropanecarboxylic acid
Lieferant: Apollo Scientific
Synonyme: 1-(3,4-Dimethoxyphenyl)cyclopropanecarboxylic Acid
OR307749-1G OR307749-500MG
APOSOR307749-1GEA 284 EUR
APOSOR307749-1G APOSOR307749-500MG
1-(3,4-Dimethoxyphenyl)cyclopropanecarboxylic acid
1-(3,4-Dimethoxyphenyl)cyclopropanecarboxylic acid
Formel: C₁₂H₁₄O₄
Molecular Weight: 222,237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08442257
CAS-Nummer: 862821-16-7