Methyl 3-aminobenzofuran-2-carboxylate 97%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-amino-2-benzo[b]furancarboxylate

OR307758-1G OR307758-5G
APOSOR307758-1GEA 71.2 EUR
APOSOR307758-1G APOSOR307758-5G
Methyl 3-aminobenzofuran-2-carboxylate 97%
Methyl 3-aminobenzofuran-2-carboxylate
Formel: C₁₀H₉NO₃
Molecular Weight: 191,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00466275
CAS-Nummer: 57805-85-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR