4-(4-Aminophenylamino)- piperidine-1-carboxylic acid benzyl ester hydrochloride
Lieferant: Apollo Scientific
OR307788-1G
OR307788-250MG
APOSOR307788-1GEA
532
EUR
APOSOR307788-1G
APOSOR307788-250MG
4-(4-Aminophenylamino)- piperidine-1-carboxylic acid benzyl ester hydrochloride
4-(4-Aminophenylamino)- piperidine-1-carboxylic acid benzyl ester hydrochloride
Formel:
C₁₉H₂₃N₃O₂·ClH Molecular Weight: 361,866 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12026442 CAS-Nummer: 1204810-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...