4-Oxo-cyclopentane-trans-1,2-dicarboxylic acid dimethyl ester

Lieferant: Apollo Scientific

OR307798-1G OR307798-5G
APOSOR307798-1GEA 0 EUR
APOSOR307798-1G APOSOR307798-5G
4-Oxo-cyclopentane-trans-1,2-dicarboxylic acid dimethyl ester
4-Oxo-cyclopentane-trans-1,2-dicarboxylic acid dimethyl ester
Formel: C₉H₁₂O₅
Lagertemperatur: Raumtemperatur
MDL: MFCD06409224
CAS-Nummer: 28269-03-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR