2-(2-(((9H-Fluoren-9-yl)methoxy)carbonylamino)ethoxy)acetic acid 98%

Lieferant: Apollo Scientific

Synonyme: 2-(2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)ethoxy)acetic acid, Fmoc-AEA, Fmoc-NH-PEG1-CH2COOH

OR307806-1G OR307806-5G
APOSOR307806-1GEA 79.9 EUR
APOSOR307806-1G APOSOR307806-5G
2-(2-(((9H-Fluoren-9-yl)methoxy)carbonylamino)ethoxy)acetic acid 98%
2-(2-(((9H-Fluoren-9-yl)methoxy)carbonylamino)ethoxy)acetic acid
Formel: C₁₉H₁₉NO₅
Molecular Weight: 341.358 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06796876
CAS-Nummer: 260367-12-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR