7-Bromo-2-chloroquinoline-3-carboxaldehyde
Lieferant: Apollo Scientific
OR307818-1G
OR307818-5G
APOSOR307818-1GEA
301
EUR
APOSOR307818-1G
APOSOR307818-5G
7-Bromo-2-chloroquinoline-3-carboxaldehyde
7-Bromo-2-chloroquinoline-3-carboxaldehyde
Formel:
C₁₀H₅BrClNO Molecular Weight: 270.51 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06239277 CAS-Nummer: 136812-31-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...