6-Ethyl-2-methylquinoline-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR307845-250MG
OR307845-100MG
APOSOR307845-250MGEA
134
EUR
APOSOR307845-250MG
APOSOR307845-100MG
6-Ethyl-2-methylquinoline-3-carboxylic acid
6-Ethyl-2-methylquinoline-3-carboxylic acid
Formel:
C₁₃H₁₃NO₂ Molecular Weight: 215,248 g/mol Schmelzpunkt: 181…182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08437576 CAS-Nummer: 92513-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...