7,8-Dimethyl-4-hydroxyquinoline

Lieferant: Apollo Scientific

OR307847-1G OR307847-5G
APOSOR307847-1GEA 293 EUR
APOSOR307847-1G APOSOR307847-5G
7,8-Dimethyl-4-hydroxyquinoline
7,8-Dimethyl-4-hydroxyquinoline
Formel: C₁₁H₁₁NO
Molecular Weight: 173.211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00272459
CAS-Nummer: 178984-37-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR