4-Amino-6-bromo-2-methylquinoline

Lieferant: Apollo Scientific

OR307857-1G OR307857-5G
APOSOR307857-1GEA 317 EUR
APOSOR307857-1G APOSOR307857-5G
4-Amino-6-bromo-2-methylquinoline
4-Amino-6-bromo-2-methylquinoline
Formel: C₁₀H₉BrN₂
Molecular Weight: 237,096 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00814272
CAS-Nummer: 96938-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR