2,6-Dichloro-8-methylquinoline-3-carboxaldehyde
Lieferant: Apollo Scientific
OR307886-1G
OR307886-5G
APOSOR307886-1GEA
301
EUR
APOSOR307886-1G
APOSOR307886-5G
2,6-Dichloro-8-methylquinoline-3-carboxaldehyde
2,6-Dichloro-8-methylquinoline-3-carboxaldehyde
Formel:
C₁₁H₇Cl₂NO Molecular Weight: 240,085 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09702833 CAS-Nummer: 938138-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...