Sodium toluene-4-sulphinate dihydrate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Toluenesulphinic acid sodium salt dihydrate
OR30790-100G
OR30790-250G
APOSOR30790-100GEA
46.4
EUR
APOSOR30790-100G
APOSOR30790-250G
Sodium toluene-4-sulphinate dihydrate 95%
Sodium toluene-4-sulphinate dihydrate
Formel:
C₇H₇O₂S·Na·2H₂O Molecular Weight: 214,215 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149642 CAS-Nummer: 7257-26-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...