2-Chloro-6-ethoxyquinoline-3-carboxaldehyde

Lieferant: Apollo Scientific

OR307920-1G OR307920-5G
APOSOR307920-1GEA 293 EUR
APOSOR307920-1G APOSOR307920-5G
2-Chloro-6-ethoxyquinoline-3-carboxaldehyde
2-Chloro-6-ethoxyquinoline-3-carboxaldehyde
Formel: C₁₂H₁₀ClNO₂
Molecular Weight: 235,666 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01250040
CAS-Nummer: 281208-98-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR