2-Chloro-6-ethylquinoline-3-carboxaldehyde
Lieferant: Apollo Scientific
OR307922-1G
OR307922-5G
APOSOR307922-1GEA
293
EUR
APOSOR307922-1G
APOSOR307922-5G
2-Chloro-6-ethylquinoline-3-carboxaldehyde
2-Chloro-6-ethylquinoline-3-carboxaldehyde
Formel:
C₁₂H₁₀ClNO Molecular Weight: 219.667 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02227042 CAS-Nummer: 436088-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...