4-Amino-7-chloro-2-methylquinoline
Lieferant: Apollo Scientific
|
Achtung
|
OR307950-1G
OR307950-250MG
APOSOR307950-1GEA
348
EUR
APOSOR307950-1G
APOSOR307950-250MG
4-Amino-7-chloro-2-methylquinoline
4-Amino-7-chloro-2-methylquinoline
Formel:
C₁₀H₉ClN₂ Molecular Weight: 192.645 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01870540 CAS-Nummer: 68017-47-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...