4-Hydrazino-6-methoxy-2-methylquinoline hydrochloride
Lieferant: Apollo Scientific
OR307974-1G OR307974-5G
APOSOR307974-1GEA 317 EUR
APOSOR307974-1G APOSOR307974-5G
4-Hydrazino-6-methoxy-2-methylquinoline hydrochloride
4-Hydrazino-6-methoxy-2-methylquinoline hydrochloride
Formel: C₁₁H₁₃N₃O·ClH
Molecular Weight: 239,701 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09997977
CAS-Nummer: 1171611-95-2