4-Hydroxy-8-methoxyquinoline-3-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

OR307980-1G
APOSOR307980-1GEA 423 EUR
APOSOR307980-1G
4-Hydroxy-8-methoxyquinoline-3-carboxylic acid 95%
4-Hydroxy-8-methoxyquinoline-3-carboxylic acid
Formel: C₁₁H₉NO₄
Molecular Weight: 219,193 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00024006
CAS-Nummer: 28027-18-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR