5-Chloro-2,8-dimethylquinoline-3-carboxylic acid ethyl ester
Lieferant: Apollo Scientific
OR307993-1G OR307993-5G
APOSOR307993-1GEA 293 EUR
APOSOR307993-1G APOSOR307993-5G
5-Chloro-2,8-dimethylquinoline-3-carboxylic acid ethyl ester
5-Chloro-2,8-dimethylquinoline-3-carboxylic acid ethyl ester
Formel: C₁₄H₁₄ClNO₂
Molecular Weight: 263.719 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787783
CAS-Nummer: 948294-10-8