6-(Hydroxymethyl)-1H-indazole

Lieferant: Apollo Scientific
Achtung

Synonyme: (1H-Indazol-6-yl)methanol

OR30859-100MG OR30859-250MG
APOSOR30859-100MGEA 21.9 EUR
APOSOR30859-100MG APOSOR30859-250MG
6-(Hydroxymethyl)-1H-indazole
6-(Hydroxymethyl)-1H-indazole
Formel: C₈H₈N₂O
Molecular Weight: 148,162 g/mol
Schmelzpunkt: 159 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11052639
CAS-Nummer: 916902-55-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR