6-(Hydroxymethyl)-1-methyl-1H-indazole 97%

Lieferant: Apollo Scientific

Synonyme: (1-Methyl-1H-indazol-6-yl)methanol

OR30863-1G OR30863-250MG
APOSOR30863-1GEA 104 EUR
APOSOR30863-1G APOSOR30863-250MG
6-(Hydroxymethyl)-1-methyl-1H-indazole 97%
6-(Hydroxymethyl)-1-methyl-1H-indazole
Formel: C₉H₁₀N₂O
Molecular Weight: 162,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11052638
CAS-Nummer: 1092961-10-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR