4-(Aminomethyl)-1H-indazole

Lieferant: Apollo Scientific
Achtung

Synonyme: (1H-Indazol-4-yl)methanamine, (1H-Indazol-4-yl)methylamine

OR30869-250MG
APOSOR30869-250MGEA 134 EUR
APOSOR30869-250MG
4-(Aminomethyl)-1H-indazole
4-(Aminomethyl)-1H-indazole
Formel: C₈H₉N₃
Molecular Weight: 147,177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10696781
CAS-Nummer: 944898-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR