7-(Aminomethyl)-1H-indazole

Lieferant: Apollo Scientific
Gefahr

Synonyme: (1H-Indazol-7-yl)methylamine

OR30870-250MG OR30870-500MG
APOSOR30870-250MGEA 149 EUR
APOSOR30870-250MG APOSOR30870-500MG
7-(Aminomethyl)-1H-indazole
7-(Aminomethyl)-1H-indazole
Formel: C₈H₉N₃
Molecular Weight: 147.177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10696780
CAS-Nummer: 944904-20-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR