6-(Aminomethyl)-1-methyl-1H-indazole hydrochloride
Lieferant: Apollo Scientific
Achtung
Synonyme: (1-Methyl-1H-indazol-6-yl)methylamine hydrochloride, (1-Methyl-1H-indazol-6-yl)methanamine hydrochloride
OR30873-100MG OR30873-500MG
APOSOR30873-100MGEA 125 EUR
APOSOR30873-100MG APOSOR30873-500MG
6-(Aminomethyl)-1-methyl-1H-indazole hydrochloride
6-(Aminomethyl)-1-methyl-1H-indazole hydrochloride
Formel: C₉H₁₁N₃·ClH
Molecular Weight: 197.665 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18632562
CAS-Nummer: 1357945-57-3