6-Acetyl-1H-indazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(1H-Indazol-6-yl)ethan-1-one, 1-(1H-Indazol-6-yl)ethanone

OR30886-1G OR30886-250MG
APOSOR30886-1GEA 393 EUR
APOSOR30886-1G APOSOR30886-250MG
6-Acetyl-1H-indazole
6-Acetyl-1H-indazole
Formel: C₉H₈N₂O
Molecular Weight: 160.173 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11869764
CAS-Nummer: 189559-85-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR