7-Acetyl-1H-indazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(1H-Indazol-7-yl)ethanone, 1-(1H-Indazol-7-yl)ethan-1-one

OR30887-1G OR30887-250MG
APOSOR30887-1GEA 58 EUR
APOSOR30887-1G APOSOR30887-250MG
7-Acetyl-1H-indazole
7-Acetyl-1H-indazole
Formel: C₉H₈N₂O
Molecular Weight: 160.173 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11869765
CAS-Nummer: 1159511-22-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR