7-Acetyl-1-methyl-1H-indazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(1-Methyl-1H-indazol-7-yl)ethanone, 7-Ethanoyl-1-methyl-1H-indazole, 1-(1-Methyl-1H-indazol-7-yl)ethan-1-one

OR30891-100MG OR30891-500MG
APOSOR30891-100MGEA 132 EUR
APOSOR30891-100MG APOSOR30891-500MG
7-Acetyl-1-methyl-1H-indazole
7-Acetyl-1-methyl-1H-indazole
Formel: C₁₀H₁₀N₂O
Molecular Weight: 174.199 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11869769
CAS-Nummer: 1159511-26-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR