6-Acetyl-2-methyl-2H-indazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Ethanoyl-2-methyl-2H-indazole, 1-(2-Methyl-2H-indazol-6-yl)ethan-1-one

OR30894-1G OR30894-250MG OR30894-500MG
APOSOR30894-1GEA 533 EUR
APOSOR30894-1G APOSOR30894-250MG APOSOR30894-500MG
6-Acetyl-2-methyl-2H-indazole
6-Acetyl-2-methyl-2H-indazole
Formel: C₁₀H₁₀N₂O
Molecular Weight: 174.199 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11869772
CAS-Nummer: 1159511-29-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR