3-Amino-3-hydroxymethyloxetane
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(3-Aminooxetan-3-yl)methanol
OR309090-500MG
OR309090-250MG
APOSOR309090-500MGEA
261
EUR
APOSOR309090-500MG
APOSOR309090-250MG
3-Amino-3-hydroxymethyloxetane
3-Amino-3-hydroxymethyloxetane
Formel:
C₄H₉NO₂ Molecular Weight: 103,12 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD20926150 CAS-Nummer: 1305208-37-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...