6-Chloro-2-methylquinoline-3-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

OR309208-1G OR309208-5G
APOSOR309208-1GEA 293 EUR
APOSOR309208-1G APOSOR309208-5G
6-Chloro-2-methylquinoline-3-carboxylic acid ethyl ester
6-Chloro-2-methylquinoline-3-carboxylic acid ethyl ester
Formel: C₁₃H₁₂ClNO₂
Molecular Weight: 249.693 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787817
CAS-Nummer: 114858-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR