7-Chloro-2,8-dimethylquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

OR309224-1G OR309224-5G
APOSOR309224-1GEA 301 EUR
APOSOR309224-1G APOSOR309224-5G
7-Chloro-2,8-dimethylquinoline-3-carboxylic acid
7-Chloro-2,8-dimethylquinoline-3-carboxylic acid
Formel: C₁₂H₁₀ClNO₂
Molecular Weight: 235,666 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787853
CAS-Nummer: 948290-22-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR