7-Methoxy-2-methylquinoline-3-carboxylic acid
Lieferant: Apollo Scientific
OR309228-1G OR309228-5G
APOSOR309228-1GEA 259 EUR
APOSOR309228-1G APOSOR309228-5G
7-Methoxy-2-methylquinoline-3-carboxylic acid
7-Methoxy-2-methylquinoline-3-carboxylic acid
Formel: C₁₂H₁₁NO₃
Molecular Weight: 217,221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03085776
CAS-Nummer: 470702-34-2