8-Ethyl-2-methylquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

OR309236-1G OR309236-5G
APOSOR309236-1GEA 325 EUR
APOSOR309236-1G APOSOR309236-5G
8-Ethyl-2-methylquinoline-3-carboxylic acid
8-Ethyl-2-methylquinoline-3-carboxylic acid
Formel: C₁₃H₁₃NO₂
Molecular Weight: 215,248 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787885
CAS-Nummer: 948291-31-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR