8-Ethylquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

OR309239-1G OR309239-5G
APOSOR309239-1GEA 310 EUR
APOSOR309239-1G APOSOR309239-5G
8-Ethylquinoline-3-carboxylic acid
8-Ethylquinoline-3-carboxylic acid
Formel: C₁₂H₁₁NO₂
Molecular Weight: 201.221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787890
CAS-Nummer: 71082-56-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR