2-Chloro-8-methylquinoline-3-methanol

Lieferant: Apollo Scientific

OR309268-1G OR309268-5G
APOSOR309268-1GEA 235 EUR
APOSOR309268-1G APOSOR309268-5G
2-Chloro-8-methylquinoline-3-methanol
2-Chloro-8-methylquinoline-3-methanol
Formel: C₁₁H₁₀ClNO
Molecular Weight: 207.656 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02232325
CAS-Nummer: 333408-31-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR