4-Hydroxy-8-methyl-2-phenylquinoline

Lieferant: Apollo Scientific

OR309299-1G OR309299-5G
APOSOR309299-1GEA 301 EUR
APOSOR309299-1G APOSOR309299-5G
4-Hydroxy-8-methyl-2-phenylquinoline
4-Hydroxy-8-methyl-2-phenylquinoline
Formel: C₁₆H₁₃NO
Molecular Weight: 235,281 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02929096
CAS-Nummer: 92855-38-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR