4-Hydroxyquinoline-6-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Carboxy-4-hydroxy-1-azanaphthalene
OR309340-1G
OR309340-250MG
APOSOR309340-1GEA
364
EUR
APOSOR309340-1G
APOSOR309340-250MG
4-Hydroxyquinoline-6-carboxylic acid
4-Hydroxyquinoline-6-carboxylic acid
Formel:
C₁₀H₇NO₃ Molecular Weight: 189.167 g/mol Schmelzpunkt: >280 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD16987424 CAS-Nummer: 1065092-81-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...