cis-2-Amino-1-cyclohexanecarboxylic acid hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: cis-2-Aminocyclohexanecarboxylic acid hydrochloride

OR309358-1G OR309358-250MG
APOSOR309358-1GEA 310 EUR
APOSOR309358-1G APOSOR309358-250MG
cis-2-Amino-1-cyclohexanecarboxylic acid hydrochloride
cis-2-Amino-1-cyclohexanecarboxylic acid hydrochloride
Formel: C₇H₁₃NO₂·ClH
Molecular Weight: 179,645 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863556
CAS-Nummer: 57266-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR