Ethyl quinazoline-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR309392-1G
OR309392-250MG
APOSOR309392-1GEA
496
EUR
APOSOR309392-1G
APOSOR309392-250MG
Ethyl quinazoline-2-carboxylate
Ethyl quinazoline-2-carboxylate
Formel:
C₁₁H₁₀N₂O₂ Molecular Weight: 202,209 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11870787 CAS-Nummer: 869299-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...