4,6-Dichloro-quinazoline-2-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

OR309397-1G OR309397-100MG OR309397-250MG OR309397-5G
APOSOR309397-1GEA 244 EUR
APOSOR309397-1G APOSOR309397-100MG APOSOR309397-250MG APOSOR309397-5G
4,6-Dichloro-quinazoline-2-carboxylic acid ethyl ester
4,6-Dichloro-quinazoline-2-carboxylic acid ethyl ester
Formel: C₁₁H₈Cl₂N₂O₂
Molecular Weight: 271,099 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10697897
CAS-Nummer: 150449-99-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR