(S)-Methyl 2,3-diaminopropanoate dihydrochloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR309421-1G
OR309421-250MG
APOSOR309421-1GEA
71.2
EUR
APOSOR309421-1G
APOSOR309421-250MG
(S)-Methyl 2,3-diaminopropanoate dihydrochloride
(S)-Methyl 2,3-diaminopropanoate dihydrochloride
Formel:
C₄H₁₀N₂O₂·2ClH Molecular Weight: 191,056 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09998031 CAS-Nummer: 147857-43-0 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...